Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SPON2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11064R-CY7
BOSSBS-11064R-CY7EA 900 EUR InStock
BOSSBS-11064R-CY7
Anti-SPON2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The Thrombospondin proteins, Thrombospondins 1-4 and Thrombospondin 5 (also designated COMP), compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix signaling. These extracellular, cell-surface proteins form complexes of both homo- and heteromultimers. Spondin-2, or Mindin, is also designated DIL-1 for its differential expression in cancerous and non-cancerous lung cells. Full-length SPON2 cDNA encodes a 331 amino acid protein with a domain arrangement similar to zebrafish F-Spondin and Mindin-1/Mindin-2: an FS1 domain, an FS2 domain, a hydrophobic signal sequence in the N-terminus and a Thrombospondin type I repeat. Immunoblot analysis demonstrates expression of dimers and oligomers in a concentration-dependent manner under nonreducing conditions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPON2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge