Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-COL2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11069R-A350
BOSSBS-11069R-A350EA 893 EUR InStock
BOSSBS-11069R-A350
Anti-COL2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Collagen type II is the most abundant protein of cartilage matrix. Two isoforms exist and collagen type IIA is involved in early development of cartilage in embryos. The collagen is produced from type II procollagen when two teloterminal propeptides are digested from the precursor and released into the circulation. Procollagen type II N-terminal propeptide is considered a marker of cartilage synthesis and the normal concentration in serum is approximately 30ng/mL.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: procollagen type IIA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge