Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-JPH3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11083R-HRP
BOSSBS-11083R-HRPEA 900 EUR InStock
BOSSBS-11083R-HRP
Anti-JPH3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Junctophilins are components of the junctional complexes between plasma membranes and endoplasmic or sarcoplasmic reticulums present in all excitable cells. Junctophilins contain a cytoplasmic domain which binds to the plasma membrane, as well as an ER/SR membrane spanning hydrophobic C-terminal segment. The three subtypes in this family are Junctophilin-1, -2 and -3. Junctophilin-1 is predominantly expressed in skeletal muscle, but is also expressed at low levels in heart. Junctophilin-2 is expressed in heart and skeletal muscle. Mutant mice lacking the Jph2 gene exhibit embryonic lethality and possess cardiac myocytes that express abnormal calcium transients. Junctophilin-3 is expressed in brain. The JPH3 alternatively spliced exon 2A has been suggested as a site for CTG repeat expansion leading to a Huntington disease-like autosomal dominant disorder.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: JPH3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge