Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LSAMP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11091R-A680
BOSSBS-11091R-A680EA 890 EUR InStock
BOSSBS-11091R-A680
Anti-LSAMP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
LSAMP is a neuronal surface glycoprotein found in cortical and subcortical regions of the limbic system expressed on the surface of somata and proximal dendrites of neurons. It is a member of the immunoglobulin (Ig) superfamily and belongs to the IgLON subfamily of cell adhesion molecules. LSAMP contains three Ig domains and a glycosylphosphatidylinositol anchor. It is a highly conserved protein between rodents and humans (99% sequence identity) and it is involved in the regulation of neurite outgrowth and mediation of proper circuit formation of limbic pathways. This suggests that LSAMP plays an important role in the development and function of the limbic system. In addition, LSAMP may function as a tumour suppressor in renal carcinomas.

Type: Primary
Antigen: LSAMP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/338
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge