Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CHRM3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11098R-A647
BOSSBS-11098R-A647EA 893 EUR InStock
BOSSBS-11098R-A647
Anti-CHRM3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Neuroligins are a family of plasma membrane proteins that possess an N-terminal hydrophobic domain, a large esterase homology domain, a single transmembrane region, a short cytoplasmic domain, and an EF-hand binding domain (1,2). Members of the neuroligin family include Neuroligin 1, Neuroligin 2 and Neuroligin 3. Neuroligins are expressed in excitatory neuronal synaptic clefts. Neuroligins play a role in the formation and remodeling of CNS synapses by binding to b-neurexins, a family of neuronal cell surface proteins. Neuroexin 1b binds to the EF-hand domain of Neuroligin 1 and requires calcium ion. Neuroligins also bind to PSD-95, which may recruit ion channels and neurotransmitter receptors to the synapses.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHRM3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge