Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Netrin G1/G2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11103R-HRP
BOSSBS-11103R-HRPEA 900 EUR InStock
BOSSBS-11103R-HRP
Anti-Netrin G1/G2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Netrin proteins are a family of laminin-related secreted proteins that provide guidance signals for axonal growth and cell migration during development. Netrin-1, which is the mammalian homolog of UNC-6 from C. elegans, is largely expressed in the developing nervous system and in mesodermal tissues. Netrin-1 is expressed by the floor plate as either a cell associated protein or in a diffusible form, and it binds to several surface receptor components, including deleted in colorectal cancer (DCC) and neogenin. During embryonic development, netrin-1 diffuses through the neuronal epithelium, where it forms a chemoattractant gradient that directs axonal migration to the ventral midline of the spinal cord. Netrin-2 and the corresponding mouse homolog netrin-3 are expressed primarily in the lower two-thirds of the spinal cord, and, like netrin-1, they can either attract or repel commissural axons at a distance. Netrin signaling is dependent on the concentration of calcium outside the cell and the level of PKA activity. In axonal cells, a reduction in PKA activity converts the responsiveness of the axons to the netrin proteins, as the cells are repelled, rather than attracted, by the netrin gradient.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Netrin G1/G2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge