Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11111R-A488
BOSSBS-11111R-A488EA 893 EUR InStock
BOSSBS-11111R-A488
Anti-PCDH1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Protocadherins are a subfamily of cadherins, a large group of related glycoproteins that mediate calcium-dependent cell-to-cell adhesion via a homophilic mechanism. Involved in a variety of functions, protocadherins help to regulate neural development and synapse formation. PCDH1 is a 1,026 amino acid single-pass type I membrane protein that contains seven cadherin domains and is a member of the protocadherin family. Localized to cell-cell and cell-matrix boundaries and expressed at high levels in brain and neuro-glial cells, PCDH1 is thought to be involved in cell adhesion and cell-cell interactions and may play a role in neuronal development. PCDH1 contains a C-terminal cytoplasmic region, an extracellular region and a transmembrane region, and is expressed as two isoforms due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge