Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDH20 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11113R-FITC
BOSSBS-11113R-FITCEA 900 EUR InStock
BOSSBS-11113R-FITC
Anti-PCDH20 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
As a subfamily of the cadherin superfamily, protocadherins are cadherin-like cell adhesion proteins that contain up to seven extracellular domains and are predominantly expressed in the nervous system. Importantly, the adhesion mechanism of protocadherins is distinct from classic cadherins. Through inactivation or overexpression, several protocadherins have been implicated in a variety of cancers. Protocadherin-20 (PCDH20), also known as protocadherin-13, is a 924 amino acid protein containing 6 cadherin domains and potentially functioning as a calcium-dependent cell-adhesion protein. In non-small cell lung cancer cell lines, a homozygous loss of PCDH20 was identified through either deletion of one allele and methylation of the other or methylation of both alleles. Hypermethylation of PCDH20 is associated with worse prognosis and clinical outcome, suggesting that PCDH20 may function as a tumor suppressor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDH20
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge