Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDHGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11155R-A350
BOSSBS-11155R-A350EA 893 EUR InStock
BOSSBS-11155R-A350
Anti-PCDHGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. The protocadherein gamma cluster consists of three subfamilies (A, B and C). As a member of the gamma subfamily B, PCDHGB1 (Protocadherin gamma B1) is a 927 amino acid protein that is one of 22 proteins encoded by the protocadherin gamma cluster. Typical of gamma protocadherins, PCDHGB1 contains six cadherin motifs and is a type I transmembrane receptor expressed in the central nervous system. With localization to synapses, members of the gamma cluster of protocadherins are essential for neuronal survival. There are two isoforms of PCDHGB1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDHGB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge