Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDHGC4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11160R-A555
BOSSBS-11160R-A555EA 1350 EUR InStock
BOSSBS-11160R-A555
Anti-PCDHGC4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHGC4 (protocadherin gamma subfamily C, 4) is a 938 amino acid single-pass type I membrane protein that localizes to the cell membrane and contains six cadherin domains. Expressed as two alternatively spliced isoforms, PCDHGC4 functions as a potential calcium-dependent cell adhesion protein that is thought to be involved in the establishment and maintenance of neuronal connections within the brain. The gene encoding PCDHGC4 maps to a protocadherin gamma gene cluster which is localized to chromosome 5 and contains over 22 protocadherin genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDHGC4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge