Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCDHGC5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11161R-FITC
BOSSBS-11161R-FITCEA 900 EUR InStock
BOSSBS-11161R-FITC
Anti-PCDHGC5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
PCDHGC5 (protocadherin gamma-C5) is a 944 amino acid single-pass type I membrane protein that contains six cadherin domains and exists as two alternatively spliced isoforms. As a potential calcium-dependent cell-adhesion protein, PCDHGC5 may be involved in the establishment and maintenance of specific neuronal connections in the brain. The gene that encodes PCDHGC5 consists of approximately 23,739 bases and maps to human chromosome 5q31.3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDHGC5
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge