Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SDK1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11164R-CY5
BOSSBS-11164R-CY5EA 900 EUR InStock
BOSSBS-11164R-CY5
Anti-SDK1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Cell adhesion molecules influence cell growth, differentiation, embryogenesis, immune response and cancer metastasis by networking information from the extracellular matrix to the cell. Sidekick-1 (SDK1) is a 2,213 amino acid single-pass membrane protein that functions as a cell adhesion molecule by guiding axonal terminals to specific synapses in developing neurons. Existing as three alternatively spliced isoforms, Sidekick-1 is expressed in retinal neurons and contains thirteen fibronectin type-III domains and six Ig-like C2-type (immunoglobulin-like) domains. Sidekick-1 expression is upregulated in glomeruli of patients with HIV-associated nephropathy, where it leads to podocyte dysfunction. The gene encoding Sidekick-1 maps to human chromosome 7p22.2 and murine chromosome 5 G2.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDK1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge