Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NARF Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11173R-CY7
BOSSBS-11173R-CY7EA 900 EUR InStock
BOSSBS-11173R-CY7
Anti-NARF Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Prenylation and methylation are two forms of protein modification, both of which are important for a variety of functions, including membrane attachment, protein-protein interactions and signaling events. NARF (nuclear prelamin A recognition factor), also known as IOP2, is a 456 amino acid nuclear protein that belongs to the NARF family. Expressed ubiquitously with highest expression in heart, skeletal muscle and brain, NARF binds to the C-terminal end of prenylated prelamin A and may be a member of a prelamin A-containing endoprotease complex. Additionally, via its association with prelamin A, NARF may be involved in heterochromatin organization. NARF is expressed as three isoforms due to alternative splicing events and, upon DNA damage, may be phosphorylated by ATM or ATR.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NARF
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge