Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KY Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11174R-FITC
BOSSBS-11174R-FITCEA 900 EUR InStock
BOSSBS-11174R-FITC
Anti-KY Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
KY peptidase (Kyphoscoliosis peptidase) is a 561 amino acid cytoskeleton protease that interacts with several sarcomeric cytoskeletal proteins, including Filamin 2. KY peptidase probably plays a role in the maturation, function and stabilization of the neuromuscular junction. KY-null mouse mutants exhibit distinct irregular subceullular Filamin 2 localization, suggesting that KY peptidase deficiency may be the cause of several types of limb-girdle muscular dystrophies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KY
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge