Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SELE Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11175R-FITC
BOSSBS-11175R-FITCEA 876 EUR InStock
BOSSBS-11175R-FITC
Anti-SELE Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Lubricin, also designated proteoglycan-4 or megakaryocyte stimulating factor, is important for boundary lubrication within articulating joints. It is a disulfide-linked homodimer (between Cysteine 1146 and Cysteine 1403) that is essential for protein cleavage. Lubricin inhibits synovial cell adhesion to the cartilage surface, but also prevents the deposition of proteins from synovial fluid onto cartilage. Lubricin is highly expressed in cartilage, liver and synovial tissue. Defects in the gene encoding for lubricin can cause Jakobs syndrome, also designated camptodactyly-arthropathy-coxa vara-pericarditis syndrome (CACP). CACP is an autosomal recessive disorder characterized by joint failure associated with noninflammatory synoviocyte hyperplasia and subinitimal fibrosis of the synovial capsule. Lubricin undergoes different levels of glycosylation and may be detected at varying molecular weights.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SELE
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge