Anti-Trypsin Inhibitor Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-11177R-A555
BOSSBS-11177R-A555EA 893 EUR InStock
BOSSBS-11177R-A555
Anti-Trypsin Inhibitor Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Cysteine-rich secretory proteins (CRISPs) represent a family of evolutionarily conserved proteins which play a role in the innate immune system and are transcriptionally regulated by androgens in several tissues. CRISP-8 (Cysteine-rich secretory protein 8), also known as PI15 (Peptidase inhibitor 15), P25TI or SugarCrisp, is a 258 amino acid secreted protein that belongs to the CRISP family. Expressed at low levels in thyroid, prostate, salivary and mammary tissue, CRISP-8 functions as a serine protease inhibitor that exhibits weak inhibitory action against Trypsin, a serine protease foundMore Product Information
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..