Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ACL6B/BAF53b Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11195R-A647
BOSSBS-11195R-A647EA 893 EUR InStock
BOSSBS-11195R-A647
Anti-ACL6B/BAF53b Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
BAF53b is a member of a family of actin-related proteins (ARPs), which shares significant sequence identity with conventional actins. Both actins and ARPs comprise an actin fold, which forms an ATP binding cleft. BAF53b is a component of the BAF (BRG/brm-associated factor) complex in mammals, which is functionally related to the SWI/SNF complex in S. cerevisiae and Drosophila, and is associated with chromatin remodelling to facilitate transcriptional activation. As neural progenitors exit the cell cycle, the highly homologous BAF53a subunit of the BAF complex is replaced by BAF53b. This subunit switch is essential for the transition from from neural stem cells / progenitors to postmitotic neurons. BAF53b also plays a role in dendritic outgrowth, during which it is required to recruit the BAF complex and the dendritic regulator CREST to specific gene promoters. This function can not be performed by BAF53a.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACL6B/BAF53b
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge