Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NR2E1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11202R-A555
BOSSBS-11202R-A555EA 893 EUR InStock
BOSSBS-11202R-A555
Anti-NR2E1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
NR2 proteins are a large family of nuclear hormone receptor transcription factors. The proteins belonging to this family are characterized by discrete domains functioning in DNA and ligand binding. NR2E1 (nuclear receptor subfamily 2, group E, member 1), also known as TLX, is an essential component in the formation of synaptic plasticity and dendritic structure in retinal astrocytes. In addition, NR2E1 is a orphan receptor that binds DNA as part of the hormone response element (HRE), a transcription regulator for hormones. DNA-binding orphan receptors have the conserved sequence 5'-AAGGTCA-3', a motif that determines substrate binding specificity. NR2E1 is expressed in brain tissue, with highest levels in astrocytes, and is localized to the nucleus. Mutations in the gene that encodes NR2E1 may lead to retinal dystrophy, a disorder characterized by a reduction in the thickness of the retina.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NR2E1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge