Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LRRTM1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11213R-A647
BOSSBS-11213R-A647EA 893 EUR InStock
BOSSBS-11213R-A647
Anti-LRRTM1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic Alpha/Beta horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. LRRTM1 (leucine rich repeat transmembrane neuronal 1) is a 522 amino acid single-pass type I membrane protein that localizes to the endoplasmic reticulum and contains ten LRR repeats. Expressed predominately in forebrain tissue, LRRTM1 is thought to be involved in the development of forebrain structures, specifically by influencing axon trafficking, as well as neuronal differentiation and connectivity. Human LRRTM1 shares 96% amino acid identity with its mouse counterpart, suggesting a conserved role between species. Defects in the gene encoding LRRTM1 may be associated with the pathogenesis of several common neurodevelopmental disorders.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRTM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge