Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SH3TC2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11221R-CY5
BOSSBS-11221R-CY5EA 900 EUR InStock
BOSSBS-11221R-CY5
Anti-SH3TC2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
SH3TC2 (SH3 domain and tetratricopeptide repeats 2) is a 1,288 amino acid protein that contains one SH3 domain and eight TPR repeats. The SH3TC2 gene encodes a protein expressed in Schwann cells of peripheral nerves, and localized to the plasma membrane and to the perinuclear endocytic recycling compartment, suggesting a possible function in myelination and/or in regions of axoglial interactions. The SH3TC2 protein is expressed in adult heart, testis, spinal cord, and brain as well as in fetal brain and liver. Mild mononeuropathy of the median nerve (MNMN) is caused by heterozygous mutation in the SH3TC2 gene. Also, Charcot-Marie-Tooth disease type 4C (CMT4C) is a more severe neuropathy caused by homozygous or compound heterozygous mutation in the SH3TC2 gene. Existing as four alternatively spliced isoforms and containing 18 exons, the SH3TC2 gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken and zebrafish, and maps to human chromosome 5q32.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SH3TC2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge