Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATRIP Ser68+Ser72 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11224R-A680
BOSSBS-11224R-A680EA 917 EUR InStock
BOSSBS-11224R-A680
Anti-ATRIP Ser68+Ser72 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
DNA damage or incomplete replication of DNA results in the inhibition of cell cycle progression at the G1 to S or the G2 to M phase transition by conserved regulatory mechanisms known as cell cycle checkpoints. Checkpoint proteins include Rad17, which is involved in regulating cell cycle progression at the G1 checkpoint as well as Chk1, Chk2, Rad1, Rad9 and Hus1, which are involved in regulating cell cycle arrest at the G2 checkpoint. In response to DNA damage, ATM and ATR kinases are important for cell cycle checkpoint response Signalling. ATR-interacting protein (ATRIP), also designated ATM and Rad3-related-interacting protein, is required for checkpoint Signalling after DNA damage. It is also important for ATR expression, which regulates DNA replication and damage checkpoint responses. ATRIP is a ubiquitously expressed protein that can form heterodimers with ATR. After dimerization they bind the RPA complex and are recruited to single stranded DNA. ATRIP is a nuclear protein that may also play a role in protein stabilisation.

Type: Primary
Antigen: ATRIP Ser68+Ser72
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge