Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-OTUB Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11229R-A350
BOSSBS-11229R-A350EA 893 EUR InStock
BOSSBS-11229R-A350
Anti-OTUB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
OTUB1 is a 271 amino acid protein that contains one OTU (ovarian tumor) domain and belongs to the OTU family of predicted cysteine proteases. Expressed as two isoforms (one of which is present throughout the body and the other of which is present only in lymphoid tissues), OTUB1 functions as a hydrolase that can remove ubiquitin residues from target proteins, thereby preventing protein degradation and playing an important role in protein turnover. OTUB1 interacts with GRAIL and, via this interaction, plays a role in the regulation and the induction of T-cell anergy, a phenomenon that occurs when T-cells are rendered unresponsive to their cognate antigens. Due to its interaction with GRAIL, OTUB1 is an important regulator of immune responses in secondary lymphoid organs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OTUB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge