Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZBTB17 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11234R
BOSSBS-11234REA 704 EUR InStock
BOSSBS-11234R
Anti-ZBTB17 Rabbit Polyclonal Antibody
Antikörper
The Myc family, including c-Myc-, N-Myc- and L-Myc, are nuclear proteins with relatively short half lives that contribute an important role in cellular processes such as proliferation, differentiation, apoptosis and transformation. The c-Myc protein activates transcription as part of a heteromeric complex with a number of interacting partners, including Max and Mxi 1; however the transforming properties of the Myc proto-oncogene are believed to be associated with Myc-mediated transcriptional repression. A POZ domain Zn finger protein, designated Miz-1 for Myc-interacting Zn finger protein-1, is a specific target of Myc-induced gene repression. Miz-1 interacts with Myc, but not Max or other Myc partners, and binding of Myc to Miz-1 requires the helix-loop-helix domain of Myc and a short amphipathic helix located in the carboxy-terminus of Miz-1. Miz-1 associates with DNA elements on the adenovirus major late and cyclin D1 promoters and activates transcription of both promoters. Expression of Miz-1 induces potent growth arrest function, and this latency is reversed by the addition of Myc.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ZBTB17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge