Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LEFTY1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11236R-A647
BOSSBS-11236R-A647EA 893 EUR InStock
BOSSBS-11236R-A647
Anti-LEFTY1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
During vertebrate embryogenesis, a left right axis is established. Secreted growth factors of the TGF beta family, including gene products derived from nodal, lefty 1 and lefty 2, play crucial roles in establishing left right asymmetries. TGF beta (Transforming growth factor beta) is a pleiotropic cytokine that regulates growth and differentiation of diverse types of cells. TGF beta actions are directed by ligand induced activation of TGF beta receptors. Complexes formed move into the nucleus, where they act as components of a transcriptional complex. Lefty, a novel member of the TGF beta superfamily, inhibits TGF beta signaling. Lefty acts to inhibit phosphorylation of Smad2 following activation of the TGF beta receptor. Lefty also inhibits events downstream from R Smad phosphorylation. Lefty provides a repressed state of TGF beta responsive genes. The Lefty family is comprised of Lefty 1 and Lefty 2 in mouse, and Lefty A and Lefty B in humans. Members of the TGF beta superfamily require processing for their activation. Cleavage is therefore an essential step for Lefty activation. Lefty is synthesized as a large inactive precursor (41 Kda) that must be endoproteolytically processed to release the bioactive polypeptide (28 kDa and 34 kDa forms). The 28kDa form induces MAPK activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LEFTY1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge