Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AP2M1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11241R-CY7
BOSSBS-11241R-CY7EA 900 EUR InStock
BOSSBS-11241R-CY7
Anti-AP2M1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Adaptins are heterotetrameric subunits of adaptors, which are complexes involved in the formation of Clathrin-coated pits for vesicle-mediated endocytosis. Clathrin and its associated heterotetrameric protein complexes make up the main protein components of the coat surrounding the cytoplasmic face of coated vesicles. The Adaptin family, comprising a, b, and g classes, is also responsible for the transport of ligand-receptor complexes from plasma membranes and the trans-Golgi network to lysosomes. Two main types of adaptor proteins (APs), AP-1 and AP-2, are found in Clathrin-coated structures located at the Golgi complex and the plasma membrane of mammalian cells, respectively. Adaptor protein complex 2 (AP-2) is composed of two large Adaptins (a1A/AP2A1 and b1/AP2B1), a medium Adaptin (m2/AP-2m1) and a small Adaptin (s2 long/AP2S1). AP-2m1, a 435 amino acid protein, links Clathrin to receptors in coated vesicles.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AP2M1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge