Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EPS15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11251R-FITC
BOSSBS-11251R-FITCEA 900 EUR InStock
BOSSBS-11251R-FITC
Anti-EPS15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Elucidation of the mechanism by which receptor tyrosine kinases (RTKs) modulate cellular physiology in response to stimuli is critical to the understanding of growth regulation. Miscues in RTK signaling pathways can result in cellular transformation and ultimately in cancer. Two novel EGF receptor substrates designated EGF-receptor pathway substrates 8 and 15, or Eps8 and Eps15, have been described. Eps8 and Eps15 are proteins, respectively that become tyrosine phosphorylated subsequent to EGF stimulation. Overexpression of Eps15 in NIH/3T3 cells causes cellular transformation, implying involvement in the regulation of cell proliferation. Eps15 is capable of binding the amino terminal portion of Crk via a conserved proline-rich domain, characteristic of all Crk binding proteins (5). Overexpression of Eps8 in both fibroblasts and hematopoietic cells results in an increased mitogenic response to EGF. Eps8 has been shown to associate with the EGF receptor despite its lack of a functional SH2 domain. Further characterization suggests the protein has both a PH domain and a SH3 domain, the functional significance of which are not yet known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPS15
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge