Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNIP1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11260R-CY7
BOSSBS-11260R-CY7EA 900 EUR InStock
BOSSBS-11260R-CY7
Anti-SNIP1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Members of the transforming growth factor-beta (TGF-Beta) superfamily play critical roles in controlling cell growth and differentiation. Effects of TGF-Beta family ligands are mediated by Smad proteins. The Smad nuclear interacting protein (SNIP1) contains a forkhead-associated (FHA) domain and acts as a nuclear inhibitor of CBP/p300. SNIP1 potently inhibits the activity of NF-kappa B, which binds the C/H1 domain of CBP/p300, by competing for the binding site. SNIP1 is also thought to induce expression of Cyclin D1 to promote cellular proliferation. SNIP1 is ubiquitously expressed with high expression in heart and skeletal muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNIP1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge