Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Syntaxin 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11261R-A750
BOSSBS-11261R-A750EA 890 EUR InStock
BOSSBS-11261R-A750
Anti-Syntaxin 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Synaptin 6 is a 255 amino acid protein that is widely expressed, with higher expression levels in brain, lung and kidney. This synaptin co-localizes with vesicle associated membrane protein (VAMP) 4 to tubular and vesicular membranes of the Golgi apparatus. The cytosolic domain of Syntaxin 6 reduces the rate on Glut4 reinternalisation upon insulin withdrawl and is involved in a memrane-trafficking process that removes Glut4 from traffic directed to the plasma membrane. Syntaxin 6 is upregulated in activated macrophages in conjunction with an increase in the secretion of cytokines. The delivery of microdomain-associated lipids and proteins to the cell surface is regulated by Syntaxin 6.

Type: Primary
Antigen: Syntaxin 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 161-255/255
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge