Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TMED10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11262R-FITC
BOSSBS-11262R-FITCEA 900 EUR InStock
BOSSBS-11262R-FITC
Anti-TMED10 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
MP21 is a member of the EMP24/GP25L/p24 cargo family of proteins that regulates vesicular trafficking in the early secretory pathway. TMP21 is a ubiquitously expressed single-pass type I membrane protein localizing to the Golgi cisternae and the plasma membrane. It contains one GOLD (Golgi dynamics) domain and participates in protein transport and quality control between the endoplasmic reticulum (ER) and the Golgi complex. In addition, TMP21 is a component of the heteromeric secretase complex (or Presenilin complex) and functions to regulate the ?secretase activity. Two isoforms, namely TMP21-I and TMP21-II, exist for this protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge