Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C1QBP/GC1q R Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11263R-CY7
BOSSBS-11263R-CY7EA 900 EUR InStock
BOSSBS-11263R-CY7
Anti-C1QBP/GC1q R Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The human complement subcomponent C1q associates with C1r and C1s in order to yield the first component of the serum complement system (SCS). The SCS contains over 30 glycoproteins that influence physiological mechanisms of the body in response to immune complex (the classical pathway), carbohydrate (the lectin pathway) or bacterial (alternative pathway) initiation. C1q binding protein (C1QBP), also designated gC1q-R, p32 (p33) or HABP1 (hyaluronan-binding protein 1), is known to bind the globular heads of C1q molecules and inhibit C1 activation. C1QBP has been described as a complement receptor for C1q on B cells, neutrophils and mast cells. The C1QBP protein may form homodimers. C1QBP is expressed in vascular endothelial cells and has been found to be a multifunctional protein interacting with elements of complement, coagulation and kinin systems. In addition, C1QBP is a subunit of pre-mRNA splicing factor SF2/ASF.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C1QBP/GC1q R
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge