Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCT2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11267R-FITC
BOSSBS-11267R-FITCEA 900 EUR InStock
BOSSBS-11267R-FITC
Anti-CCT2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
CCT2 is one of eight largely unrelated subunit proteins found in a protein chaperone complex known as the chaperonin-containing TCP-1 (CCT) or TRiC complex. The CCT complex is an abundanct cytoslic component that is credited with helping newly synthesized polypeptides adopt the correct conformation (1). Proteins that fold and assemble with the help of CCT include the cytoskeletal proteins actin and tubulin as well as up to 15% of newly synthesized eukaryotic proteins (2). CCT2 is the β-subunit of the chaperone complex and is one of several CCT proteins that exhibit increased expression in response to stress. This implies that the CCT complex helps cells recover from protein damage by assisting in protein folding and assembly (3). CCT subunit levels also change throughout the cell cycle, with lower proteins levels (and reduced chaperone activity) found during induced cell cycle arrest during at M phase (4). Each CCT subunit is thought to perform a specific function during protein folding and assembly (5); CCT2 exhibits both actin and tubulin binding activities (6,3) but the exact molecular function on this subunit remains uncertain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCT2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge