Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HSP56/FKBP52 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11270R-A680
BOSSBS-11270R-A680EA 890 EUR InStock
BOSSBS-11270R-A680
Anti-HSP56/FKBP52 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
HSP56 is a cis-trans prolyl isomerase belonging to the immunophilin protein family. The human HSP 56 gene (FKBP4) has multiple polyadenylation sites and the HSP 56 protein can undergo phosphorylation. HSP 56 influences immunoregulatory gene expression in lymphocytes, protein folding and trafficking. It can serve as a co-chaperone for steroid hormone nuclear receptors to govern appropriate hormone action in target tissues. The protein can associate with phytanoyl-CoA alpha-hydroxylase (PHYH) and with HSP90 through a series of tetratricopeptide repeat (TPR) domains. HSP 56 is a TRPC ion channel accessory protein that modulates channel activation following receptor stimulation.

Type: Primary
Antigen: HSP56/FKBP52
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/459
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge