Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-XCL1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11287R-CY7
BOSSBS-11287R-CY7EA 900 EUR InStock
BOSSBS-11287R-CY7
Anti-XCL1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Lymphotactin is a chemokine-like molecule produced selectively, if not exclusively, by activated CD8+ T cells (1). Lymphotactin is a C-type member of the chemokine superfamily, but retains only the 2nd and 4th of the four cysteine residues conserved in other chemokines (3,4). In normal tissues, lymphotactin is expressed at high levels in spleen, thymus, small intestine, and peripheral blood leukocytes, as well as at low levels in lung, prostate, and ovary (2). Lymphotactin is chemotactic for CD4+ and CD8+ T cells but not for monocytes, and induces a rise in intracellular calcium in peripheral blood lymphocytes (1). The specific receptor for lymphotactin is XCR1, a member of the G protein-coupled receptor family (5). The gene which encodes lymphotactin maps to human chromosome 1q23 (1,2).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: XCL1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge