Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HSD17B4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11296R-CY7
BOSSBS-11296R-CY7EA 883 EUR InStock
BOSSBS-11296R-CY7
Anti-HSD17B4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
17Beta-HSD4 (17Beta-hydroxysteroid dehydrogenase type 4) is also known as peroxisomal multifunctional enzyme/protein 2 (MFE-2/MFP-2), D-bifunctional enzyme or 17-Beta Estradiol dehydrogenase type IV. It belongs to the 17Beta-HSD family of proteins that regulate the availability of steroids within various tissues throughout the body. 17Beta-HSD4 inactivates Estradiol through its oxidative activity but it is primarily involved in peroxisomal fatty acid and cholesterol Beta-oxidation. It has a multi-domain structure: the dehydrogenase domain is fused to a hydratase and a lipid transfer domain. 17Beta-HSD4 is a target protein of chromeceptin and it is essential for the downstream activation of Stat6. 17Beta-HSD4-deficient patients exhibit Zellweger-like syndrome and die within the first year of life. They display neuronal migration defects, facial dysmorphisms, severe hypotonia and convulsions in the neonatal period.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSD17B4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge