Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IL2RB Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11300R-CY7
BOSSBS-11300R-CY7EA 929 EUR InStock
BOSSBS-11300R-CY7
Anti-IL2RB Rabbit Polyclonal Antibody (Cy7®)
Antikörper
IL2 Receptor beta (CD122) is a member of the immunoglobulin superfamily that forms the high affinity IL2 receptor with CD25 and CD132. This receptor chain, which is also shared by the IL15 receptor, is constitutively expressed by NK cells and at lower levels by T cells, B cells, monocytes, and macrophages. The IL2 Receptor beta chain can combine with either the common gamma subunit (gamma c) alone or the gamma c subunit and the IL2 Receptor alpha subunit to generate intermediate or high affinity IL2 receptor complexes, respectively. CD122 levels can be upregulated by activation stimuli such as IL2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL2RB
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge