Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCYT1A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11306R-A680
BOSSBS-11306R-A680EA 890 EUR InStock
BOSSBS-11306R-A680
Anti-PCYT1A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Increase in fetal surfactant synthesis and lung maturity is caused by the glucocorticoidal induction of enzymes required for phosphatidylcholine synthesis towards the end of gestation. The regulation of gestational age-dependent induction of phosphatidylcholine synthesis by glucocorticoids is still unclear. The rate-controlling enzyme in the phosphatidylcholine biosynthetic pathway is CTP-phosphocholine cytidylyltransferase A (CCT A). In cultured eukaryotic cells, this enzyme is essential for survival. The alpha isoform is located in the nucleus and is regulated by reversible phosphorylation and membrane association. There is significant identity between the alpha-helical membrane-binding domains of CCT A and soybean oleosin. Expressed CCT A has lipid-dependent cytidylyltransferase activity. The gene which encodes CCT A maps to human chromosome 3q.

Type: Primary
Antigen: PCYT1A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/367
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge