Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SAP102 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11315R-CY3
BOSSBS-11315R-CY3EA 900 EUR InStock
BOSSBS-11315R-CY3
Anti-SAP102 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
MPP3, also known as Discs large homolog 3 (DLG3), is a 585 amino acid protein belonging to the MAGUK family. Proteins in the MAGUK family, including CASK, ZO-1, ZO-2, p55 and MPP2, are known to regulate cell proliferation, intracellular junctions and signaling pathways by interacting with the cytoskeleton. MPP3 contains one guanylate kinase-like domain, one PDZ domain, two L27 domains and one SH3 domain. Via the PDZ domain, MMP3 interacts with the C-terminus of TSLC1. MMP3 has also been found to interact with SR-2A and SR-4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SAP102
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge