Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PSD95BP3 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11316R-CY3
BOSSBS-11316R-CY3EA 900 EUR InStock
BOSSBS-11316R-CY3
Anti-PSD95BP3 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Members of the postsynaptic density-95 (PSD-95)/SAP90 family of membrane-associated guanylate kinase (MAGUK) proteins function as multimodular scaffolds that organize protein-signaling complexes at neuronal synapses. SAPAP3 (PSD-95/SAP90-binding protein 3), also known as DLGAP3 (disks large-associated protein 3) or DAP3, is a 979 amino acid protein that belongs to the SAPAP family of PSD-95/SAP90-associated proteins. Localized to the cell junction, as well as to the peripheral membrane and the cell synapse, SAPAP3 is thought to play a role in the organization of neuronal cell signaling and synapses, and may also function as an adaptor protein, possibly linking ion channels with the cytoskeleton.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSD95BP3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge