Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TMEM259 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11322R-FITC
BOSSBS-11322R-FITCEA 900 EUR InStock
BOSSBS-11322R-FITC
Anti-TMEM259 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The gene encoding human Membralin, C19orf6, localizes to chromosome 19p13.3. It contains 11 exons, which encode at least two splice variants in human cancer. Membralin is a multi-pass membrane protein and exists either as a long or short form. The long form of Membralin comprises all 11 exons, while the short form contains all exons except exon 10. Expression of different Membralin isoforms depends on tissue type. The long form is expressed in ovarian and colorectal carcinomas, whereas the short form is expressed in breast or pancreatic carcinomas. Because Membralin expression in ovarian carcinomas is highest in serous carcinomas as compared to surface epithelium carcinomas, expression of Membralin may be useful as a novel tumor-associated marker in ovarian serous carcinomas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TMEM259
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge