Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PTPN5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11328R-A555
BOSSBS-11328R-A555EA 893 EUR InStock
BOSSBS-11328R-A555
Anti-PTPN5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The brain-specific STEP (striatal enriched phosphatase) family of protein tyrosine phosphatases (PTPs) comprises both transmembrane and cytosolic protein members which are the products of alternative splicing. STEP family members are expressed in the dopaminoceptive neurons of the CNS, with highest expression in the basal ganglia and related structures. The STEP protein regulates the N-methyl-d-aspartate receptor (NMDAR) complex; STEP depresses both NMDAR single-channel activity and synaptic currents. The membrane-associated STEP61 isoform localizes in the postsynaptic densities (PSDs) of striatal neurons. STEP61 contains a single tyrosine phosphatase domain, two proline-rich domains and two transmembrane domains. The STEP61 protein associates with the Src family kinase member Fyn when Fyn is phosphorylated at Tyr-420 and not Tyr-431. Upon association, STEP61 dephosphorylates Tyr-420 residue and may thus regulate Fyn activity in PSDs. Isolated from mouse brain, the STEP20 isoform lacks the conserved tyrosine phosphatase domain. The human STEP gene maps to chromosome 11p15.2-p15.1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPN5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge