Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SORCS1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11330R-CY5
BOSSBS-11330R-CY5EA 900 EUR InStock
BOSSBS-11330R-CY5
Anti-SORCS1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
There are three sorCS genes that have diverse, partially overlapping functions in the central nervous system. In the developing and mature central nervous system, the homologous SorCS1 and SorCS2 genes and the SorCS3 gene are expressed in a combinatorial, non-overlapping pattern. SorCS proteins show homology to the mosaic receptor SorLA and the neurotensin receptor sortilin, based on a common VPS10 domain, which is the hallmark of the SorCS receptor family. SorCS1 is a type 1 receptor containing a VPS10P domain and a leucine-rich domain. Alternative splicing of human SorCS1 results in four isoforms with different cytoplasmic tails and differential expression in tissues. Human SorCS1 is detected in fetal and infant brain and in fetal retina. Alternative splicing of murine SorCS1 also results in four isoforms. Murine isoform 1 is highly expressed in brain and at lower levels in heart, liver and kidney. It is detected in newborn mouse brain and in adult olfactory bulb and cerebral cortex. Murine isoform 2 is highly expressed in liver and at lower levels in heart, brain, kidney and testis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SORCS1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge