Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNAP91 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11333R-A350
BOSSBS-11333R-A350EA 893 EUR InStock
BOSSBS-11333R-A350
Anti-SNAP91 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Clathrin-coated pits and vesicles are assembled for receptor-mediated endocytosis through interaction with Clathrin associated protein complexes. Vesicle transport is mediated from the trans-Golgi network by the adapter complex AP-1 and from the plasma membrane by the AP-2 complex. The AP-1 and AP-2 adapter protein complexes consist of Clathrin binding Adaptin proteins (g and b1 for AP-1, a and b2 for AP-2) and two smaller subunits known as AP50 and AP17. The a and b Adaptin chains have a similar two-domain organization with C-terminal domains that vary in both sequence and length. a-Adaptin splice variants A and C display variable relative expression levels and differential distribution in different tissues. AP-3 (also designated AP180 or F1-20) is a synapse-specific Clathrin assembly protein. The protein CALM (Clathrin assembly protein lymphoid myeloid leukemia) is highly homologous to AP180 and may also be involved in Clathrin assembly.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNAP91
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge