Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLSTN2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11339R-A555
BOSSBS-11339R-A555EA 893 EUR InStock
BOSSBS-11339R-A555
Anti-CLSTN2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Members of the calsyntenin protein family are localized to the post-synaptic membrane of exicitatory central nervous system (CNS) synapses. Calsyntenin-2, also known as Alcadein-gamma, is a 955 amino acid protein that localizes to the endoplasmic reticulum, golgi apparatus and plasma membranes. Containing 2 cadherin-like repeats in its N-terminal extracellular region, calsyntenin-2 binds synaptic calcium with its cytoplasmic domain, suggesting a role in the modulation of calcium-mediated postsynaptic signals. Under normal physiological conditions, calsyntenin-2 is protoeolytically processed in an event in which the primary zeta-cleavage generates a short C-terminal transmembrane fragment and a long extracellular N-terminal domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLSTN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge