Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11340R-A350
BOSSBS-11340R-A350EA 893 EUR InStock
BOSSBS-11340R-A350
Anti-CPEB3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The regulated translation of messenger RNA is essential for cell-cycle progression, establishment of the body plan during early development and modulation of key activities in the central nervous system. Cytoplasmic polyadenylation, one mechanism of controlling translation, is driven by cytoplasmic polyadenylation element binding proteins, called CPEBs. CPEB3 (cytoplasmic polyadenylation element binding protein 3) is a 698 amino acid protein that contains two RNA recognition motif (RRM) domains and, like other CPEB proteins, may play a role in the maturation of the central nervous system. CPEB3 exists as multiple alternatively spliced isoforms that are encoded by a gene which maps to human chromosome 10, which houses over 1,200 genes and comprises nearly 4.5% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPEB3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge