Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CPLX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11341R-A350
BOSSBS-11341R-A350EA 893 EUR InStock
BOSSBS-11341R-A350
Anti-CPLX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Complexin 1 and Complexin 2, also designated Synaphin 1 and Synaphin 2, contain an a-helical middle domain of approximately 58 amino acids. Complexin 1 and Complexin 2 are expressed in presynaptic terminals of inhibitory and excitatory hippocampal neurons, respectively, and in cytoplasmic pools during early stages of development. Complexins promote SNARE (soluble N-ethylmaleimide-sensitive factor attachment protein receptors) precomplex formation by binding to synaxin with its a-helical domain. Complexins are important regulators of transmitter release at a late step in calcium dependent neurotransmitter release or immediately after the calcium-triggering step of fast synchronous transmitter release and preceding vesicle fusion. Neurons lacking complexins show reduced transmitter release efficiency due to decreased calcium sensitivity of the synaptic secretion process. Complexin 2 may play a role in LTP (long term potentiation) following tetanic stimulation. A progressive loss of Complexin 2 occurs in the brains of mice carrying the Huntington disease mutation, an autosomal dominant neurodegenerative disorder. Changes in the neurotransmitter release might contribute to the motor, emotional and cognitive dysfunctions seen in these mice.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPLX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge