Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ICA1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11349R-A555
BOSSBS-11349R-A555EA 893 EUR InStock
BOSSBS-11349R-A555
Anti-ICA1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Carbonic anhydrases (CAs), also designated carbonate dehydratases or carbonate hydrolyases, form a large family of genes that encode zinc metalloenzymes of great physiologic importance. As catalysts of the reversible hydration of carbon dioxide, these enzymes participate in a variety of biologic processes, including respiration, acid-base balance, bone resorption and calcification as well as the formation of aqueous humor, cerebrospinal fluid, saliva and gastric acid. Genes in the ?carbonic anhydrase family encode either active carbonic anhydrase isozymes or 揳catalytic?(devoid of CO2 hydration activity) carbonic anhydrase-related proteins. Human CA I (CA1) is encoded by the CA1 gene, which maps to a region on chromosome 8 that harbors a cluster of CA genes. CA I localizes to the cytoplasm and research indicates that a severe deficiency of CA I does not result in any obvious hematological or renal consequences.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ICA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge