Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Piccolo Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11354R-HRP
BOSSBS-11354R-HRPEA 1350 EUR InStock
BOSSBS-11354R-HRP
Anti-Piccolo Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Piccolo is 1 component of the presynaptic cytomatrix. Bassoon is a large protein which is also found in the presynaptic compartments of rat brain synapses. Bassoon, along with Piccolo, is part of the ensemble of presynaptic proteins that are involved in orchestrating events at the nerve terminal. Bassoon is found in axon terminals of hippocampal neurons where it is highly concentrated in the vicinity of the active zone. Piccolo has a similar distribution and colocalizes with Bassoon in cultured hippocampal cells. Piccolo zinc fingers interact with the dual prenylated rab3A and VAMP2/ Synaptobrevin II receptor PRA. Bassoon may be involved in cytomatrix organization at the site of neurotransmitter release. The gene which encodes bassoon maps to human chromosome 3p21.31. The gene which encodes piccolo maps to human chromosome 7q21.11.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Piccolo
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge