Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RIC8A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11355R-HRP
BOSSBS-11355R-HRPEA 900 EUR InStock
BOSSBS-11355R-HRP
Anti-RIC8A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The Ras superfamily of GTPases can be subdivided into the Ras, Rho/Rac, Sar, Rab, Arf, Rap and Ran subfamilies, all of which control multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. The Ras superfamily of GTPases function as regulated switches that toggle between a biologically active GTP-bound and an inactive GDP-bound form. This activation is catalyzed by guanine nucleotide exchange factors (GEFs). RIC-8A (resistance to inhibitors of cholinesterase 8 homolog A), also known as RIC8 or Synembryn-A, is a 530 amino acid cytoplasmic protein that can activate several G-alpha proteins, including G?i-1, G?q and G?o. Functioning as a guanine nucleotide exchange factor, RIC-8A binds to GDP-associated substrates and exchanges bound GDP for free GTP. Via its ability to stimulate protein function, RIC-8A plays a role in regulating mitotic movement and may be involved in receptor-mediated ERK activation. RIC-8A is expressed as three isoforms due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RIC8A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge