Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RIMS3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11359R-FITC
BOSSBS-11359R-FITCEA 900 EUR InStock
BOSSBS-11359R-FITC
Anti-RIMS3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Rab 3, a neural/neuroendocrine-specific member of the Rab family, is involved in Ca2+-regulated exocytosis and functions in an inhibitory capacity, controlling the recruitment and pooling of secretory vesicles at the plasma membrane. The Rim (Rab 3 interacting molecule) family of proteins (Rim1, Rim2, Rim3 and Rim4) are multidomain adaptors that regulate Rab 3 activity and sub-sequent neurotransmitter release. Rim3, also known as RIMS3 (regulating synaptic membrane exocytosis 3) or NIM3, is a 308 amino acid member of the Rim family. Localized to the synapse and to cell junctions, Rim3 contains one C2 domain and is thought to play an important role in the regulation of synaptic membrane exocytosis. Rim3, a protein that may be phosphorylated upon DNA damage, is expressed throughout the body with highest levels present in brain tissue.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RIMS3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge