Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RIMS4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11360R-FITC
BOSSBS-11360R-FITCEA 900 EUR InStock
BOSSBS-11360R-FITC
Anti-RIMS4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Rim4 (Rab 3 interacting molecule 4), also known as Rim4 or regulating synaptic membrane exocytosis protein 4, is a 269 amino acid protein that localizes to the cell junction and regulates synaptic membrane exocytosis.Rab 3, a neural/neuroendocrine-specific member of the Rab family, is involved in Ca2+-regulated exocytosis. Rab 3 functions in an inhibitory capacity by controlling the recruitment of secretory vesicles into a releasable pool at the plasma membrane. Rim (Rab 3 interacting molecule), a putative effector protein for Rab 3 proteins, is thought to regulate neutrotransmitter release through its interaction with Rab 3 and other synaptic proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RIMS4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge